Learn More
Medchemexpress LLC Mind4-19 | 129544-85-0 | 99.93% | 373.47 | 50 MG
SDP
Catalog No. 5000361244
Encompass_Preferred
MIND4-19 is a potent SIRT2 inhibitor with an IC50 value of 7.0 μM. It can be used for researching Huntington's disease.
  • Potent SIRT2 inhibitor.
  • Exhibits an IC50 value of 7.0 μM for SIRT2.
  • Applicable for research into Huntington's disease.
Catalog No. 50-003-61244 Supplier Medchemexpress LLC Supplier No. HY14723350MG
MIND4-19 is a potent SIRT2 inhibitor with an IC50 value of 7.0 μM. It can be used for researching Huntington's disease.
  • Potent SIRT2 inhibitor.
  • Exhibits an IC50 value of 7.0 μM for SIRT2.
  • Applicable for research into Huntington's disease.