Learn More
Medchemexpress LLC Mindeudesivir hydrobromide | 2779498-79-0 | 98.16% | C24H31DBrN5O7 | 50 MG
SDP
Catalog No. 5000356236
Encompass_Preferred
Mindeudesivir hydrobromide is a deuterated version of Remdesivir, a highly orally active nucleoside antiviral effective against SARS-CoV-2 and respiratory syncytial virus (RSV). It maintains the antiviral activity of Remdesivir against COVID-19 and is the first domestically produced deuterium targeting COVID-19. This product is for research use only.
  • Highly orally active nucleoside antiviral
  • Effective against SARS-CoV-2 and respiratory syncytial virus (RSV)
  • Maintains antiviral activity against COVID-19
  • First domestically produced deuterium targeting COVID-19
  • For research use only
Catalog No. 50-003-56236 Supplier Medchemexpress LLC Supplier No. HY145119AS50MG
Mindeudesivir hydrobromide is a deuterated version of Remdesivir, a highly orally active nucleoside antiviral effective against SARS-CoV-2 and respiratory syncytial virus (RSV). It maintains the antiviral activity of Remdesivir against COVID-19 and is the first domestically produced deuterium targeting COVID-19. This product is for research use only.
  • Highly orally active nucleoside antiviral
  • Effective against SARS-CoV-2 and respiratory syncytial virus (RSV)
  • Maintains antiviral activity against COVID-19
  • First domestically produced deuterium targeting COVID-19
  • For research use only