Learn More
Medchemexpress LLC Mindeudesivir hydrobromide | 2779498-79-0 | 98.2% | C24H31DBrN5O7 | 100 MG
SDP
Catalog No. 5000355630
Encompass_Preferred
Mindeudesivir hydrobromide is a deuterated version of Remdesivir, acting as a highly orally active nucleoside antiviral against SARS-CoV-2 and respiratory syncytial virus (RSV). This compound maintains the antiviral activity of Remdesivir against COVID-19.
  • Extend the half-life in vivo
  • Improve oral bioavailability
  • Improve metabolic characteristics
  • Improve drug safety
  • Preserve therapeutic properties
Catalog No. 50-003-55630 Supplier Medchemexpress LLC Supplier No. HY145119AS100MG
Mindeudesivir hydrobromide is a deuterated version of Remdesivir, acting as a highly orally active nucleoside antiviral against SARS-CoV-2 and respiratory syncytial virus (RSV). This compound maintains the antiviral activity of Remdesivir against COVID-19.
  • Extend the half-life in vivo
  • Improve oral bioavailability
  • Improve metabolic characteristics
  • Improve drug safety
  • Preserve therapeutic properties