Learn More
Medchemexpress LLC Mindeudesivir hydrobromide | 2779498-79-0 | 98.2% | C24H31DBrN5O7 | 25 MG
SDP
Catalog No. 5000357174
Encompass_Preferred
Mindeudesivir hydrobromide (VV116) is a deuterated version of Remdesivir, a highly orally active nucleoside antiviral agent against SARS-CoV-2 and respiratory syncytial virus (RSV). It maintains the antiviral activity of Remdesivir against COVID-19 and is noted as the first domestically produced deuterium targeting COVID-19. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs, potentially offering several advantages.
  • Used as a tracer or internal standard for quantitative analysis.
  • Extends in vivo half-life and improves oral bioavailability.
  • Improves metabolic characteristics and drug safety.
  • Preserves therapeutic properties.
  • Inhibits RSV replication in A549 cells.
  • Shows strong antiviral activity against RSV in mice.
  • Exhibits favorable pharmacokinetic properties and good safety profile.
Catalog No. 50-003-57174 Supplier Medchemexpress LLC Supplier No. 25MGHY145119AS
Mindeudesivir hydrobromide (VV116) is a deuterated version of Remdesivir, a highly orally active nucleoside antiviral agent against SARS-CoV-2 and respiratory syncytial virus (RSV). It maintains the antiviral activity of Remdesivir against COVID-19 and is noted as the first domestically produced deuterium targeting COVID-19. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs, potentially offering several advantages.
  • Used as a tracer or internal standard for quantitative analysis.
  • Extends in vivo half-life and improves oral bioavailability.
  • Improves metabolic characteristics and drug safety.
  • Preserves therapeutic properties.
  • Inhibits RSV replication in A549 cells.
  • Shows strong antiviral activity against RSV in mice.
  • Exhibits favorable pharmacokinetic properties and good safety profile.