Learn More
Precision Solution Inc MINEBEA SIGNUM ADVANCED SCALE
SDP
Catalog No. NC2090483
Encompass
Minebea Signum Advanced Scale Model SIWADCP-3-35-S 35 kg capacity 60,000 division high resolution 25 button keyboard 14 segment display
This item is not returnable. View return policy
Catalog No. NC2090483 Supplier Precision Solution Inc Supplier No. SIWADCP335S
Minebea Signum Advanced Scale Model SIWADCP-3-35-S 35 kg capacity 60,000 division high resolution 25 button keyboard 14 segment display