Learn More
Sigma Aldrich Fine Chemicals Biosciences Mineral oil, 8042-47-5, MFCD00131611, 500 mL
Catalog No. NC0696894
Encompass
For molecular biology, BioReagent, light oil. Mineral Oil is a colorless, odorless mixture of alkanes from a mineral source. It is often used as an inert overlay to minimize evaporation in molecular biology applications, including PCR, but has many other applications on other research fields.
Catalog No. NC0696894 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M5904500ML
For molecular biology, BioReagent, light oil. Mineral Oil is a colorless, odorless mixture of alkanes from a mineral source. It is often used as an inert overlay to minimize evaporation in molecular biology applications, including PCR, but has many other applications on other research fields.