Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Mineral Oil
SCI_ED

Catalog No. S25440 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Heavy
Light
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25440 500 mL Heavy
S25439 500 mL Light
S25439A 4 L Light
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25440
Add to Cart
Add to Cart

High-purity mineral oil for instructional lab use. Ideal for chemistry and general science classroom experiments.

  • Intended for instructional science labs and demonstrations
  • High Purity: Assay range of 99 to 100% w/v for consistent lab results
  • ACS Grade: Suitable for academic lab experiments in chemistry and biology
  • Physical Properties: Density: 1.2610g/mL at 20°C
  • Formula Weight: 92.1
  • Linear Formula: C3H5(OH)3
  • Packaging: Supplied as a 4-liter lab-grade liquid
  • Applications: Commonly used as a solvent, humectant, or for viscosity experiments
  • For educational use only, Certificate of Analysis (C of A) not available

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Density Gravity: 0.83 at 15.6°C
Assay Percent Range >99%
Packaging Heavy
Quantity 500 mL
Formula Weight Variable
Physical Form Liquid
Chemical Name or Material Mineral Oil
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.