Learn More
Ace Glass, Inc. MINERAL OIL BUBBLER MINERAL OIL BUBBLER
SDP
Catalog No. 509558581
Encompass_Preferred
Mineral oil bubbler with 8mm O.D. tubing connections is used to make a vent to the atmosphere. Unit may also be used with mercury when adequate ventilation is available. Reservoir head prevents oil from being sucked back into the system. Volume is approximately 40mL below side arm.
Catalog No. 50-955-8581 Supplier Ace Glass, Inc. Supplier No. 876110
Mineral oil bubbler with 8mm O.D. tubing connections is used to make a vent to the atmosphere. Unit may also be used with mercury when adequate ventilation is available. Reservoir head prevents oil from being sucked back into the system. Volume is approximately 40mL below side arm.