Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Oil, Fisher BioReagents

Catalog No. BP26291 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP26291 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP26291

missing translation for 'validMainframeMsg'

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

Glass Bottle; 1L

Mineral Oil can be used in PCR applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mineral Oil
Name Note Clear, Viscous Liquid
CAS 8042-47-5
Color White
Boiling Point 260°C
Packaging Glass Bottle
Molecular Formula C16H10N2Na2O7S2
Quantity 1 L
Specific Gravity 0.818 to 0.880 (at 25°C)
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
Molecular Weight (g/mol) 452.363
PubChem CID 9566064
Vapor Pressure <0.1 mmHg
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.