Learn More
Alkali Scientific Mineral Oil, Light, 20 Liters
SDP
Catalog No. 5000659687
Encompass_Preferred
Mineral Oil, Light, is a hydrophobic lubricant and carrier medium commonly used in laboratory settings. The 20-liter container provides a large volume for industrial and research applications requiring a stable and non-reactive medium. Light mineral oil is used in various experiments as a protective barrier, preventing the evaporation of volatile substances and ensuring smooth mixing in biochemical assays. It is also used in tissue culture as an overlay to protect delicate cell cultures from contamination. The oil is often used in biological research, microscopy, and to maintain the integrity of samples during long-term storage or incubation.
Catalog No. 50-006-59687 Supplier Alkali Scientific Supplier No. INE002CP
Mineral Oil, Light, is a hydrophobic lubricant and carrier medium commonly used in laboratory settings. The 20-liter container provides a large volume for industrial and research applications requiring a stable and non-reactive medium. Light mineral oil is used in various experiments as a protective barrier, preventing the evaporation of volatile substances and ensuring smooth mixing in biochemical assays. It is also used in tissue culture as an overlay to protect delicate cell cultures from contamination. The oil is often used in biological research, microscopy, and to maintain the integrity of samples during long-term storage or incubation.