Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Oil, Molecular Biology Grade, Hoefer™
SDP

Catalog No. 03500521
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-500-521 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-500-521 Supplier Hoefer, Inc Supplier No. GR1381
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IEF, 2D electrophoresis, PCR

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Quantity 1 L
Grade Molecular Biology
Physical Form Oil
Chemical Name or Material Mineral Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.