Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral oil, for spectroscopy, suitable for nujol mull preparations for IR-spect.

Catalog No. AC124020010
Change view
Click to view available options
Quantity:
6 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC124020010 1 L Glass Bottle
AC124020060 6 mL Glass Bottle
AC124020025 2.5 L Glass Bottle
AC124020100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC124020010 Supplier Thermo Scientific Chemicals Supplier No. 124020010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For IR spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00131611
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Colorless
Density 0.8510g/mL
Flash Point 185°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.4620 to 1.4730
Quantity 1 L
Specific Gravity 0.851
Merck Index 14, 7186
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, chloroform, ether,, carbon dislulfide, petroleum ether, oils, miscible with wax and fats, insoluble in alcohol
Viscosity 14-19 mm2/s (40°C)
Grade Spectroscopy
Physical Form Oily Liquid
Chemical Name or Material Mineral oil
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
EINECSNumber 232-455-8
RTECSNumber PY8047000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.