Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mineral Spirits, Spectrum™ Chemical
SDP

Catalog No. 18604629 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-629 20 L Steel Can
18-604-630 500 mL Steel Can
18-604-631 4 L Steel Can
18-604-632 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-629 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M129220LTMT
Add to Cart
Add to Cart

M1292, 64475-85-0

Spectrum™ Chemical Mineral Spirits , also known as white spirit or mineral turpentine, is a petroleum-derived liquid used as an organic solvent in decorating and painting. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 64475-85-0

Specifications

CAS Max % 1
Quantity 20 L
Packaging Steel Can
UN Number UN1268
Boiling Point 149°C to 213°C
Grade Reagent
Chemical Name or Material Mineral Spirits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.