Learn More
Research Products International Corp Mini-100 Orbital-Genie, Benchtop Orbital Shaker
SDP
Catalog No. 501537680
Encompass
Mini-100 Orbital-Genie is ideal for gentle shaking applications with no additional accessories necessary. The Mini-100 features variable shaking speeds from 20-100 rpm with a non-slip, rubber coated platform with spill-proof edges.
A customizable mixing action creates any mixing action from a smooth vortex to sloshing aeration or anything in between. The Orbital-Gene's heavy duty base has a combination of weight and specially designed feet to prevent walking even at the highest speed. Shaker is ready to use as supplied and is ideal for shaking of odd shaped vessels and containers.
Catalog No. 50-153-7680 Supplier Research Products International Corp Supplier No. 400805
Mini-100 Orbital-Genie is ideal for gentle shaking applications with no additional accessories necessary. The Mini-100 features variable shaking speeds from 20-100 rpm with a non-slip, rubber coated platform with spill-proof edges.
A customizable mixing action creates any mixing action from a smooth vortex to sloshing aeration or anything in between. The Orbital-Gene's heavy duty base has a combination of weight and specially designed feet to prevent walking even at the highest speed. Shaker is ready to use as supplied and is ideal for shaking of odd shaped vessels and containers.