Learn More
Research Products International Corp Stacking Tray for Mini-100 Orbital-Genie
SDP
Catalog No. 501537681
Encompass
A non-slip tray that can be stacked onto the Mini-100 Orbital-Genie's existing platform tray, or GTS 100's Non-Slip Platform, increasing the capacity for mixing by double-stacking two trays. The stackable tray measures 290 x 290mm (11.4 x 11.4in).
Catalog No. 50-153-7681 Supplier Research Products International Corp Supplier No. 400806
A non-slip tray that can be stacked onto the Mini-100 Orbital-Genie's existing platform tray, or GTS 100's Non-Slip Platform, increasing the capacity for mixing by double-stacking two trays. The stackable tray measures 290 x 290mm (11.4 x 11.4in).