Learn More
BioSpec Products 2.0 ml Vial Adapter for Mini-BeadBeater-16
SDP
Catalog No. NC0400963
Encompass_Preferred
2.0 ml screwcap microvial adapter for the Mini-BeadBeater-16, holds sixteen 2ml vials
Catalog No. NC0400963 Supplier BioSpec Products Supplier No. 607TC16
2.0 ml screwcap microvial adapter for the Mini-BeadBeater-16, holds sixteen 2ml vials