Learn More
REPLIGEN CORPORATION Repligen MiniKros Filter, 500 kD MWCO, 2600 cm² Surface Area, Sterile
SDP
Catalog No. NC3443168
Encompass
The Repligen MiniKros Hollow Fiber Filter is a compact, sterile ultrafiltration module designed for efficient TFF in scientific applications. It features a 500 kDa mPES membrane with exceptional flow characteristics, facilitating optimal biomolecule processing. With an effective length of 20 cm and 830 fibers, it offers a large surface area of 2,600 cm², making it ideal for scaling small to medium volume samples. Its straightforward connections enhance integration into existing systems, setting it apart from competitors.
Catalog No. NC3443168 Supplier REPLIGEN CORPORATION Supplier No. N02E50005S
The Repligen MiniKros Hollow Fiber Filter is a compact, sterile ultrafiltration module designed for efficient TFF in scientific applications. It features a 500 kDa mPES membrane with exceptional flow characteristics, facilitating optimal biomolecule processing. With an effective length of 20 cm and 830 fibers, it offers a large surface area of 2,600 cm², making it ideal for scaling small to medium volume samples. Its straightforward connections enhance integration into existing systems, setting it apart from competitors.