Learn More
Research Products International Corp Mini-300 Orbital-Genie, Adhering Mat
SDP
Catalog No. 501976890
Encompass
The Mini-300 Orbital-Genie with adhering mat provides a flat platform with a non-slip adhering rubber mat perfect for use with open containers, bottles and T-flasks. Compact design fits any lab bench and durable housing withstands years of rugged use.
Patented material is naturally adhering and rinses clean for new adhering properties. Unit allows for variable shaking speeds up to 300 RPM.
Catalog No. 50-197-6890 Supplier Research Products International Corp Supplier No. 400836
The Mini-300 Orbital-Genie with adhering mat provides a flat platform with a non-slip adhering rubber mat perfect for use with open containers, bottles and T-flasks. Compact design fits any lab bench and durable housing withstands years of rugged use.
Patented material is naturally adhering and rinses clean for new adhering properties. Unit allows for variable shaking speeds up to 300 RPM.