Learn More
Brady™ Mini All Purpose Cable Lockout
Catalog No. 19221459 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Green
Red
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-221-459 Green
19-221-458 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-221-459 Supplier Brady™ Supplier No. 179483
Add to Cart
Add to Cart
Expand lockout capabilities in tight spaces with a handle-free design that isolates multiple energy points easily and effectively.
  • Compact, handle-free profile: Fits easily into tight spaces
  • Multi-point energy isolation: Secures multiple points, tricky valve handles and complex panels at once
  • Rugged, textured finish: Ultra-durable housing with a high-grip surface finish
  • One-person application: Simple, intuitive design allows a single worker to thread, cinch and lock out quickly
  • Slip-free cable hold: High-strength pivoting arm cinches cable tight to hold firm under tension
Specifications
Product Type Cable Lockout
Color Green
Material ABS
Dimensions (D x W x H) 1.33 x 4.06 x 3.26 in.
Length (English) 3.26 in.
Quantity 1 Ea.