Learn More
BioSpec Products Mini-Beadbeater-96, 115 volt
SDP
Catalog No. NC0141170
Encompass
The MiniBeadbeater-96 disrupts cells and tissue by violently shaking a sample inside a classic 2 ml microvial or standard deep well microplate partially filled with tiny ceramic or steel beads.  Complete cell disruption is achieved in 1 to 3 minutes of beadbeating.  A common application is nucleic acid extraction.  Up to 400 mg of tissue is added to a vial containing glass or ceramic beads and a nucleic acid extraction solution...the later often being purchased as part of a nucleic acid extraction kit.  The synergy of simultaneous mechanical cell disruption and nucleic acid extraction in a chaotropic solution gives very high yields of high quality RNA or DNA.  Most protocols developed using smaller capacity models of the MBB are transferable with little if any modification.
Catalog No. NC0141170 Supplier BioSpec Products Supplier No. 1001
Add to Cart
Add to Cart
Request a Quote
The MiniBeadbeater-96 disrupts cells and tissue by violently shaking a sample inside a classic 2 ml microvial or standard deep well microplate partially filled with tiny ceramic or steel beads.  Complete cell disruption is achieved in 1 to 3 minutes of beadbeating.  A common application is nucleic acid extraction.  Up to 400 mg of tissue is added to a vial containing glass or ceramic beads and a nucleic acid extraction solution...the later often being purchased as part of a nucleic acid extraction kit.  The synergy of simultaneous mechanical cell disruption and nucleic acid extraction in a chaotropic solution gives very high yields of high quality RNA or DNA.  Most protocols developed using smaller capacity models of the MBB are transferable with little if any modification.