Learn More
Research Products International Corp Mini BeadBeater 96 Disruptor
SDP
Supplier:  Research Products International Corp 112017
Encompass
Mini-Beadbeaters feature distinct advantages over competitive models including affordability, disruption speed, recovery efficiency, proven reliability and maximum performance. Designed for high-throughput analytical screening, the Mini-BeadBeater 96, efficiently and safely disrupts samples of spores, microorganisms, plant and animal tissue or soils samples at one time. Once samples are loaded with beads and unit is turned on, they are violently shaken for up to 5 minutes - and disruption of over 95% is quickly achieved.
Catalog No. 50-197-6805
Add to Cart
Request a Quote