Learn More
Heathrow Scientific Mini Benchtop Cooler, -20°C, Blue
Description
- Ideal for keeping samples or reagents cool during testing
- Polycarbonate construction with rubber non-slip feet
- Coolers 160011 and 160012 are filled with a self-insulating gel to improve cooling capacity of unit
- Gridded and Keyed one-position lids for easy sample identification
- 0.5 mL Tube Adapters supplied as standard with the 0.5 to 2.0 mL tube size versions
Specifications
Specifications
| Dimensions (L x W x H) | 5.58 x 4.02 x 3.3 in. |
| Array | 3 x 4 |
| Holds Temp., Hours | ≤-15°C, Up to 2 hr. |
| Material | Polycarbonate |
| Product Type | Mini Cooler |
| Color | Blue |
| Holds | 12 Tubes (0.5 to 2 mL) |
| Autoclavable | Not Autoclavable |
| Temperature Control | Passive |
| Temperature Range | -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.