Learn More
Chemglass Life Sciences Mini Bio Reactor Tube, 50 mL, PP/HDPE, Sterile, Vent Cap
SDP
Catalog No. 502733514
Encompass
Mini bio reactor tubes are 50 mL conical tubes designed for small-scale suspension culture and biological reaction experiments that require aseptic conditions and gas exchange. Each tube features clear graduations and a large white marking area, and is packaged sterile for parallel screening and optimization workflows.
  • 50 mL conical volume.
  • USP VI polypropylene tube with HDPE vent cap.
  • Vent cap contains a 0.22 μm hydrophobic membrane for gas exchange.
  • Sterile; non-pyrogenic; non-cytotoxic; DNase/RNase-free; human DNA-free.
  • Easy-to-read black graduations and a large white marking area.
  • Packaged sterile in zip-sealable bags; pack quantity P10.
Catalog No. 50-273-3514 Supplier Chemglass Life Sciences Supplier No. CGE4285050
Mini bio reactor tubes are 50 mL conical tubes designed for small-scale suspension culture and biological reaction experiments that require aseptic conditions and gas exchange. Each tube features clear graduations and a large white marking area, and is packaged sterile for parallel screening and optimization workflows.
  • 50 mL conical volume.
  • USP VI polypropylene tube with HDPE vent cap.
  • Vent cap contains a 0.22 μm hydrophobic membrane for gas exchange.
  • Sterile; non-pyrogenic; non-cytotoxic; DNase/RNase-free; human DNA-free.
  • Easy-to-read black graduations and a large white marking area.
  • Packaged sterile in zip-sealable bags; pack quantity P10.