Learn More
Andwin Scientific MINI BIOMIXER W/ DIPPLED MAT
SDP
Catalog No. NC0262883
Encompass_Preferred
The 115V BioMixer Mini Rocker is platform shaker that features a 3-dimensional nutating motion for moderately aggressive mixing. It includes a dimpled mat ideal for blood and centrifuge tubes.
Catalog No. NC0262883 Supplier Andwin Scientific Supplier No. B3D1020
The 115V BioMixer Mini Rocker is platform shaker that features a 3-dimensional nutating motion for moderately aggressive mixing. It includes a dimpled mat ideal for blood and centrifuge tubes.