Learn More
Med Vet International Mini Blood Collection Tube - Brown/Serum Gel, J1565D, Each
Catalog No. 5000583778
Encompass_Preferred
Mini Tubes 1.3ml Smaller tubes for smaller patients with snap-on color coded caps and Labeled for patient info. Squared off bottom allows tube to stand upright. ?O? ring under cap for tight seal. J1565d Serum Gel. Brown.
Catalog No. 50-005-83778 Supplier Med Vet International Supplier No. JOR1565D
Mini Tubes 1.3ml Smaller tubes for smaller patients with snap-on color coded caps and Labeled for patient info. Squared off bottom allows tube to stand upright. ?O? ring under cap for tight seal. J1565d Serum Gel. Brown.