Learn More
Sigma Aldrich Fine Chemicals Biosciences Mini BlotBoyTM 3D platform1EA
Catalog No. 502905549
Encompass
The Mini BlotBoy platform rockers provide the perfect 3-dimensional motion for all blot related applications. The tilt angle and speed have been optimally set for gentle but thorough mixing in gel trays boxes and other flat vessels.Mini BlotBoys are supplied with autoclavable flat mats and are ideal for blot washes and gel staining/destaining. Designed to conserve bench space the Mini BlotBoy is supplied with a 10.5 x 7.5 inch platform. The Mini BlotBoy can accept stackable platforms handles loads up to 2kg and has maintenance free brushless motors safe for use in cold rooms and incubators.
Catalog No. 50-290-5549 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7555161EA
The Mini BlotBoy platform rockers provide the perfect 3-dimensional motion for all blot related applications. The tilt angle and speed have been optimally set for gentle but thorough mixing in gel trays boxes and other flat vessels.Mini BlotBoys are supplied with autoclavable flat mats and are ideal for blot washes and gel staining/destaining. Designed to conserve bench space the Mini BlotBoy is supplied with a 10.5 x 7.5 inch platform. The Mini BlotBoy can accept stackable platforms handles loads up to 2kg and has maintenance free brushless motors safe for use in cold rooms and incubators.