Learn More
Electron Microscopy Sciences mini-BUCK™ High Flow Calibrator, 120 V
SDP
Catalog No. 5033959
Encompass
The mini-BUCK Primary Flow Calibrator is the industry standard for day-to-day flow verification and calibration of your personal and environmental sampling pumps. It is also suitable for verifying rotameters, setting instrument panels, and calibrating laboratory instruments. No more lugging oversized calibration panels. No more rotameter inaccuracies. The mini BUCK is accurate at any attitude and can be used for flow measurements with sampling media.
Flow: 100cc/min 30LPM
Catalog No. 50-339-59 Supplier Electron Microscopy Sciences Supplier No. 6524063
The mini-BUCK Primary Flow Calibrator is the industry standard for day-to-day flow verification and calibration of your personal and environmental sampling pumps. It is also suitable for verifying rotameters, setting instrument panels, and calibrating laboratory instruments. No more lugging oversized calibration panels. No more rotameter inaccuracies. The mini BUCK is accurate at any attitude and can be used for flow measurements with sampling media.
Flow: 100cc/min 30LPM