Learn More
Fine Science Tools Mini Bulldog Serrefines, 28mm
SDP
Catalog No. NC0447934
Encompass
Product: Mini Bulldog Serrefine - Curved/28mm/ Shape: Curved/ Jaw Dimensions: 6 x 2mm/ Clamping Pressure: Medium/ Alloy: Stainless Steel/ Length: 28mm/ Serrated: Yes/ Unit: Each
Catalog No. NC0447934 Supplier Fine Science Tools Supplier No. 1805328
Product: Mini Bulldog Serrefine - Curved/28mm/ Shape: Curved/ Jaw Dimensions: 6 x 2mm/ Clamping Pressure: Medium/ Alloy: Stainless Steel/ Length: 28mm/ Serrated: Yes/ Unit: Each