Learn More
Azzota Corp Mini Centrifuge, Come With Exchangeable Rotors, One For 1.5 Ml Microtubes And One For 0.2 Ml Pcr Strip Tubes. Two Adapters, One Is 0.5 Ml And Another Is 0.2 Ml. Speed: 10000Rpm (2200Xg)
Catalog No. 507532960
Encompass_Preferred
Mini Centrifuge, Come with exchangeable rotors, one for 1.5 ml microtubes and one for 0.2 ml PCR strip tubes. Two adapters, one is 0.5 ml and another is 0.2 ml. Speed: 10000rpm (2200Xg)
Catalog No. 50-753-2960 Supplier Azzota Corp Supplier No. MINISPIN10K
Mini Centrifuge, Come with exchangeable rotors, one for 1.5 ml microtubes and one for 0.2 ml PCR strip tubes. Two adapters, one is 0.5 ml and another is 0.2 ml. Speed: 10000rpm (2200Xg)