Learn More
Labrepco Inc MyFuge Mini Centrifuge, Purple Lid, 8-Place Rotor
SDP
Catalog No. NC0903275
Encompass
The MyFuge Mini centrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, the MyFuge is extremely easy to use. Simply close the lid and the rotor instantly reaches 6,000 rpm. Open the lid, and the rotor quickly decelerates to a stop for removing samples. Specifications: Speed: 6,000 rpm / 2,000 xg Capacity: 8 x 1.5/2.0ml tubes / 16 x 0.2ml PCR Tubes / 2 x PCR strips (8 x 0.2ml) Dimensions (W x D x H): 4.5 x 5.9 x 4.5 in. / 11.4 x 15 x 11.4 cm Weight: 2.2 lbs. (1 kg) Electrical Data: Universal Voltage - 100-240V, 50-60 Hz, 250W Warranty: 1 year Lid Color: Purple Features: Eight position microtube rotor Storage compartment for PCR rotor Near silent operation Starts and stops in just seconds Conserves valuable bench space
Catalog No. NC0903275 Supplier Labrepco Inc Supplier No. C1008P
The MyFuge Mini centrifuge is a personal centrifuge that virtually fits in the palm of your hand. Ideal for quick spin downs of microtubes and PCR tubes, the MyFuge is extremely easy to use. Simply close the lid and the rotor instantly reaches 6,000 rpm. Open the lid, and the rotor quickly decelerates to a stop for removing samples. Specifications: Speed: 6,000 rpm / 2,000 xg Capacity: 8 x 1.5/2.0ml tubes / 16 x 0.2ml PCR Tubes / 2 x PCR strips (8 x 0.2ml) Dimensions (W x D x H): 4.5 x 5.9 x 4.5 in. / 11.4 x 15 x 11.4 cm Weight: 2.2 lbs. (1 kg) Electrical Data: Universal Voltage - 100-240V, 50-60 Hz, 250W Warranty: 1 year Lid Color: Purple Features: Eight position microtube rotor Storage compartment for PCR rotor Near silent operation Starts and stops in just seconds Conserves valuable bench space