Learn More
Biotang Inc TSZ mini centrifuge max speed 7000rpm, with two rotors, ideal for quick spin down and microfiltration, tiny footprint is only 4 1/2 in. (One Fixed Angle rotor for 6X0.2 (PCR)/0.5/1.5/2 ml tubes, one rotor for two strip tubes (8X0.2 strips)
Catalog No. 507514951 Shop All BIOTANG Inc Products
Encompass_Preferred
TSZ mini centrifuge max speed 7000rpm, with two rotors, ideal for quick spin down and microfiltration, tiny footprint is only 4 1/2 in. (One Fixed Angle rotor for 6X0.2 (PCR)/0.5/1.5/2 ml tubes, one rotor for two strip tubes (8X0.2 strips)
Catalog No. 50-751-4951 Supplier Biotang Inc Supplier No. TSZC02
TSZ mini centrifuge max speed 7000rpm, with two rotors, ideal for quick spin down and microfiltration, tiny footprint is only 4 1/2 in. (One Fixed Angle rotor for 6X0.2 (PCR)/0.5/1.5/2 ml tubes, one rotor for two strip tubes (8X0.2 strips)