Learn More
Medicus Health MINI CENTRIFUGE
SDP
Catalog No. NC1862873
Encompass
∙ Mini Centrifuge has an Ergonomic Design and Safety Measures, the Sprout Plus Mini Centrifuge is Easy to Use and brings increased efficiency to the lab∙Tool-free SnapSpin rotors and quick-spin options make the centrifuges fast and easy to use∙Locking lid was functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction with one hand∙Large lid size accommodates longer and skirted tubes and tall tube lids∙Braking function quickly stops spinning when powered off or when lid is opened∙Weighted rubberized base eliminates potential movement and dampens vibration and noise∙Ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1.5/2.0, 0.5, 0.2 mL) for customized use∙Specifications: Maximum Speed: Approx. 6,000 rpm (fully loaded), Maximum RCF: 2000 x g, Dimensions: 6" x 5" x 4.3", Certifications: UL (Canada and US), ROHS3, CE, WEEE and C-Tick, Five Year Warranty
Catalog No. NC1862873 Supplier Medicus Health Supplier No. 7041M1
Add to Cart
Add to Cart
∙ Mini Centrifuge has an Ergonomic Design and Safety Measures, the Sprout Plus Mini Centrifuge is Easy to Use and brings increased efficiency to the lab∙Tool-free SnapSpin rotors and quick-spin options make the centrifuges fast and easy to use∙Locking lid was functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction with one hand∙Large lid size accommodates longer and skirted tubes and tall tube lids∙Braking function quickly stops spinning when powered off or when lid is opened∙Weighted rubberized base eliminates potential movement and dampens vibration and noise∙Ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1.5/2.0, 0.5, 0.2 mL) for customized use∙Specifications: Maximum Speed: Approx. 6,000 rpm (fully loaded), Maximum RCF: 2000 x g, Dimensions: 6" x 5" x 4.3", Certifications: UL (Canada and US), ROHS3, CE, WEEE and C-Tick, Five Year Warranty