Learn More
Biotang Inc Mini-Plate Centrifuge, 500g for spin for 2 PCR plates; Quickly Spin down droplets;Accepts skirted, non-skirted and all standard PCR plates;Less than 1/4 the size of most plate centrifuges
Catalog No. 507514949 Shop All BIOTANG Inc Products
Encompass_Preferred
Mini-Plate Centrifuge, 500g for spin for 2 PCR plates; Quickly Spin down droplets;Accepts skirted, non-skirted and all standard PCR plates;Less than 1/4 the size of most plate centrifuges
Catalog No. 50-751-4949 Supplier Biotang Inc Supplier No. MINIP25
Mini-Plate Centrifuge, 500g for spin for 2 PCR plates; Quickly Spin down droplets;Accepts skirted, non-skirted and all standard PCR plates;Less than 1/4 the size of most plate centrifuges