Learn More
World Precision Instrument Mini Centrifuge, purple lid, 2 rotors, 240V
SDP
Catalog No. 502539376
Encompass
This personal centrifuge fits in the palm of your hand. It is deal for quick spin downs of either microtubes or PCR tubes.Close the lid and the rotor instantly ramps up to 6000 RPM. Open the lid and it decelerates to a stop. The 8-position microtube rotor is incuded, along with a second rotor for 0.2mL PCR tubes and strips. The second rotor is stored (with the removal key) in a compartment on the bottom of the centrifuge. 
Catalog No. 50-253-9376 Supplier World Precision Instrument Supplier No. 504328
This personal centrifuge fits in the palm of your hand. It is deal for quick spin downs of either microtubes or PCR tubes.Close the lid and the rotor instantly ramps up to 6000 RPM. Open the lid and it decelerates to a stop. The 8-position microtube rotor is incuded, along with a second rotor for 0.2mL PCR tubes and strips. The second rotor is stored (with the removal key) in a compartment on the bottom of the centrifuge. 