Learn More
Bioworld Mini-Columns, 0.5 ml (100/pk), 1 CS
Catalog No. 50198569
Encompass_Preferred
0.5 ml Microspin columns, 200/casemolded in polypropylenefeatures a HDPE filter of medium porosity (45-90 micron)
Catalog No. 50-198-569 Supplier Bioworld Supplier No. 211600001
0.5 ml Microspin columns, 200/casemolded in polypropylenefeatures a HDPE filter of medium porosity (45-90 micron)