Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences True North Mini Cooler for 32 x 0.5/2.0 mL tubes, 4 x 8 array Tubes
SDP

Catalog No. 501926825
Encompass_Preferred

Protect temperature-sensitive samples at the bench or in the freezer. Ideal for keeping PCR reagents, enzymes and other biochemical reagents cool while working at the bench. Filled with insulating gel, the Mini Coolers can also be used for controlled freezing of samples longer temperature retention. Features polycarbonate construction with rubber non-slip feet and gridded and keyed one-position lids for easy sample identification.
Retention: 1°C (7.5 hr)
Tubes: 4 x 8 array
Lid: Gel-Filled
Dimensions: 245 x 105 x 100
Temp: 0°C

Catalog No. 50-192-6825 Supplier Electron Microscopy Sciences Supplier No. 41133
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protect temperature-sensitive samples at the bench or in the freezer. Ideal for keeping PCR reagents, enzymes and other biochemical reagents cool while working at the bench. Filled with insulating gel, the Mini Coolers can also be used for controlled freezing of samples longer temperature retention. Features polycarbonate construction with rubber non-slip feet and gridded and keyed one-position lids for easy sample identification.
Retention: 1°C (7.5 hr)
Tubes: 4 x 8 array
Lid: Gel-Filled
Dimensions: 245 x 105 x 100
Temp: 0°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.