Learn More
Electron Microscopy Sciences Mini Cooling Chamber , 15 Holes, 4" x 2½" x 1½"
SDP
Catalog No. 501902297
Encompass_Preferred
Mini-cooling chambers are ideal for use when only a small number of microtubes are required. Each Mini-Chamber will fit in our 1L Glacierbrand Ice Pan(6.13" x 6.13" x 3.61"). Mini-Chambers are anodized aluminum and utilize our "Surround-A-Tube" cooling technology.
Capacity: 15 Holes
Dimensions: 4" x 2½" x 1½"
Use: 1.5-2.0 ml Tubes
Catalog No. 50-190-2297 Supplier Electron Microscopy Sciences Supplier No. 6860020
Mini-cooling chambers are ideal for use when only a small number of microtubes are required. Each Mini-Chamber will fit in our 1L Glacierbrand Ice Pan(6.13" x 6.13" x 3.61"). Mini-Chambers are anodized aluminum and utilize our "Surround-A-Tube" cooling technology.
Capacity: 15 Holes
Dimensions: 4" x 2½" x 1½"
Use: 1.5-2.0 ml Tubes