Learn More
Electron Microscopy Sciences EMS High Precision Cutting Tweezer Mini Style 152, 100 mm
SDP
Catalog No. 501930587
Encompass_Preferred
For cutting hair sprigs, electronic component wires, copper, gold, silver and other soft wires. Large head, oblique strong cut at the cutter's tip. Cutting edge is limited to the tip area. 111 mm long. M = 11 mm, N = 3 mm, P = 24 mm.
OAL: 100 mm
Catalog No. 50-193-0587 Supplier Electron Microscopy Sciences Supplier No. 78325152
For cutting hair sprigs, electronic component wires, copper, gold, silver and other soft wires. Large head, oblique strong cut at the cutter's tip. Cutting edge is limited to the tip area. 111 mm long. M = 11 mm, N = 3 mm, P = 24 mm.
OAL: 100 mm