Learn More
Research Products International Corp Mini-Desiccator Cabinet, 13 1/2 x 10 x 8 1/2 Inches
SDP
Catalog No. 501977592
Encompass
This cabinet benchtop unit is ideal for storing and protecting moisture sensitive reagents and samples. Cabinet is molded of clear polystyrene with a clear copolymer door that allows see through viewing of contents. Features a positive door latch and gasket for airtight sealing of chamber. Locking loops allow the use of a padlock or a tamper-proof seal for added security. A built-in dial hygrometer displays the interior relative humidity.
WARNING  This product can expose you to chemicals including Vinyl Chloride and Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7592 Supplier Research Products International Corp Supplier No. 420116
This cabinet benchtop unit is ideal for storing and protecting moisture sensitive reagents and samples. Cabinet is molded of clear polystyrene with a clear copolymer door that allows see through viewing of contents. Features a positive door latch and gasket for airtight sealing of chamber. Locking loops allow the use of a padlock or a tamper-proof seal for added security. A built-in dial hygrometer displays the interior relative humidity.
WARNING  This product can expose you to chemicals including Vinyl Chloride and Bisphenol-A, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.