Learn More
Sigma Aldrich Fine Chemicals Biosciences Mini Dialysis Kit 1 kDa Cy
Catalog No. 502904059
Encompass
The disposable tubes in Mini Dialysis Kit offer a simple SOLUTIONon to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.
Catalog No. 50-290-4059 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE80648375
The disposable tubes in Mini Dialysis Kit offer a simple SOLUTIONon to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.