Learn More
Sigma Aldrich Fine Chemicals Biosciences Mini DiazaldR apparatus Jo1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1088981EA
Encompass
The mini Diazald apparatus bridges the gap between the System 45 diazomethane generator Z411736 (for preparing 1 mmol of diazomethane) and the Diazald kit (for preparing ~100 mmol of diazomethane). It consists of a reaction vessel and a condenser in one compact unit. This apparatus features a cold-finger in place of a water-jacketed condenser. When filled with dry ice/acetone slush the condenser efficiently prevents dangerous diazomethane/ether vapor from escaping. A separatory funnel Z100382 and a round-bottom receiving flask Z100358 (must be equipped with Clear-Seal(R) joints) are the only other glassware needed and can be purchased separately.NOTE Sigma-Aldrich is unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
Catalog No. 50-290-8605