Learn More
Alkali Scientific MyTemp™ Mini Digital Incubator, with Heating Only, 115V
SDP
Catalog No. 502336015
Encompass_Preferred
With digital temperature control, the MyTemp™ incubators provide convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Despite a modest footprint of 14.5 x 13", the MyTemp™ Mini incubator features large internal chambers, capable of storing flasks and bottles up to 2 liters. In addition, the incubators include two adjustable/removable shelves for increased capacity.
Catalog No. 50-233-6015 Supplier Alkali Scientific Supplier No. RS7164
With digital temperature control, the MyTemp™ incubators provide convenient "set and walk away" operation, eliminating the need for external thermometers and repetitive "fine tuning" of an analog control knob. Simply choose the desired temperature and the incubator immediately begins to heat up while accurately monitoring the chamber temperature and conveniently displaying the temperature in real time on the large LED control panel. Despite a modest footprint of 14.5 x 13", the MyTemp™ Mini incubator features large internal chambers, capable of storing flasks and bottles up to 2 liters. In addition, the incubators include two adjustable/removable shelves for increased capacity.