Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mini Dino Counters
SCI_ED

Catalog No. S79001
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S79001 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79001 Supplier Learning Resources, Inc Supplier No. LER 0710
Add to Cart
Add to Cart

Includes

16 page booklet of dinosaur facts and activities.

Learning with dinosaurs

These prehistoric creatures make perfect party favors, classroom rewards or fun teaching tools! Great for counting, sorting and imaginary play.

Order Info

Soft rubber dinosaur counters are 2 in. tall and come in six shapes and colors.

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Mini Dino Counters
Includes 16 Page booklet of dinosaur facts and activities
For Use With (Application) These prehistoric creatures make perfect party favors, classroom rewards or fun teaching tools! Great for counting, sorting and imaginary play
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.