Learn More
World Precision Instrument Mini Dissecting Scissors, 9.5cm, Straight, Sharp Tips, Large Rings
SDP
Supplier:  World Precision Instrument 503245
Encompass
Our Mini Dissecting Scissors with Large Rings are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces. They are commonly used in microsurgery, like ophthalmic surgery. These dissecting scissors have large rings for people with larger fingers. These scissors are 9.5cm (3.75") long and have straight blades with rounded tips and fine blades. (Standard and medium blades are also available on our Mini Dissecting Scissors. Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel.
Catalog No. 50-253-9533