Learn More
World Precision Instrument Mini Dissecting Scissors, 9cm, Curved, Fine Tips, Left Hand
SDP
Supplier:  World Precision Instrument 503627
Encompass
Our Left-Hand Mini Dissecting Scissors are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces. They are commonly used in microsurgery, like ophthalmic surgery. These scissors are 9 cm (3.5") long and have straight blades with sharp tips or curved blades with fine tips. Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. These are designed for left handed people. These are available in surgical grade stainless steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Catalog No. 50-253-9467