Learn More
Cole-Parmer Extech EX330 Mini Autoranging MultiMeter with Temp + Voltage Detector
Catalog No. 50992601 Shop All Cole-Parmer Products
Encompass
These compact multimeters feature a built-in noncontact AC voltage detector with a red LED indicator and audible beeper ensuring operator safety. Meters provide 100 to 600 VAC measurement capability with 0.5% basic accuracy. Additional features include low-battery indicator and an easy-to-read large 1" LCD digit display. Model 20005-28 provides manual ranging capabilities with a 2000-count display. Model 20005-29 features auto-ranging capabilities with a 2000-count display. Meter includes max hold, data hold, and auto power off conserving battery life. Model 20005-30 features auto-ranging capabilities with a 4000-count display and type K thermocouple measurements (°F/°C switchable). Additional functions include capacitance, frequency, and relative function.
Catalog No. 50-992-601 Supplier Cole-Parmer Supplier No. 2000530
These compact multimeters feature a built-in noncontact AC voltage detector with a red LED indicator and audible beeper ensuring operator safety. Meters provide 100 to 600 VAC measurement capability with 0.5% basic accuracy. Additional features include low-battery indicator and an easy-to-read large 1" LCD digit display. Model 20005-28 provides manual ranging capabilities with a 2000-count display. Model 20005-29 features auto-ranging capabilities with a 2000-count display. Meter includes max hold, data hold, and auto power off conserving battery life. Model 20005-30 features auto-ranging capabilities with a 4000-count display and type K thermocouple measurements (°F/°C switchable). Additional functions include capacitance, frequency, and relative function.