Learn More
USA Scientific Inc Mini Dry Bath, 100C
Catalog No. NC0248930
Encompass
Thermal-Lok mini dry heat bath (without blocks), digital display, 5C above ambient to 100C
Catalog No. NC0248930 Supplier USA Scientific Inc Supplier No. 25201001
Thermal-Lok mini dry heat bath (without blocks), digital display, 5C above ambient to 100C