Learn More
Azzota Corp Mini Dry Bath, 0°C~100°C, RT + 5°C~100°C, L114×W140×H107. 2lbs
Catalog No. NC2330063
Encompass_Preferred
Mini Dry Bath, 0C~100C, RT + 5C~100C, L114W140H107. 2lbs.
Catalog No. NC2330063 Supplier Azzota Corp Supplier No. KDB100
Mini Dry Bath, 0C~100C, RT + 5C~100C, L114W140H107. 2lbs.