Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Mini Dry Bath with Heated Lid

Catalog No. 14955241 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Range (Metric):
Ambient +5° to 100°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
14-955-241 Ambient +5° to 100°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-955-241
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Compact dry bath block heater with heated lid to reduce condensation and evaporation.

The Fisherbrand™ Mini Dry Bath with Heated Lid features a unique heating system that improves temperature performance and reduces condensation and evaporation. With a second heating element located within the lid of the instrument, samples are also heated from the top, providing a uniform sample temperature throughout the sample.

Both the block and lid temperature can be selected independently in increments of 0.1°C, making this mini dry bath suitable for the most temperature sensitive applications, including, LAMP assays and immunoassay procedures.

Note: Bath does not include blocks. See Accessories links for block options.

Specifications

Alarm Types Visible
Autoclavable Not Autoclavable
Capacity (Metric) 15 x 1.5 mL
Chamber Material ABS Plastic
CO2 Sensor Technology No Sensor
Oxygen Control None
Temperature Range (Metric) Ambient +5° to 100°C
Type Dry Bath with Heated Lid
Includes Mini Dry Bath. (Blocks are sold separately.)
Temperature Accuracy 0.3°C
Temperature Uniformity 0.2°C
Warranty 2 Years
Dimensions (L x W x H) 6.2 x 5.3 x 4.7 in. (15.8 x 13.5 x 12 cm)
Weight (English) 1.8 lb.
Voltage 120/230 V
Amperage 1 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.