Learn More
USA Scientific Inc Mini Dry Heat Bath HC digital
Catalog No. 502744834
Encompass
Thermal-Lok Mini Dry Heat Bath HC with cooling (without blocks), digital display, 25C below ambient to 100C
Catalog No. 50-274-4834 Supplier USA Scientific Inc Supplier No. 25301001
Thermal-Lok Mini Dry Heat Bath HC with cooling (without blocks), digital display, 25C below ambient to 100C